Vivaflow® 50 Modular Tangential Flow System, Sartorius
Supplier: Avantor
CA76408-738EA 4869.69 CAD
CA76408-738
Vivaflow® 50 Modular Tangential Flow System, Sartorius
Filters Tangential Flow Systems
Vivaflow® 50 is a disposable and ready-to-use crossflow cassette for filtration and concentration of samples from 100 ml to 3 L.
  • Modular, scalable to six units
  • Optimal for aqueous sample concentration e.g. proteins and nanoparticles
  • High flow rate and recovery
The membrane surface per device is 50 cm². One pack contains two devices and comes with all necessary accessories for running two devices with a laboratory pump and a size 16 pump head.
For speeding up the concentration and increasing the volume throughput, six cassettes can be connected.
Packaging: Pump, easy load pump head (size 16), tubing, 500 ml sample/diafiltration reservoir, module stand, pressure indicator, T connectors, series interconnectors.