Sartolab® P20 and P20 Plus Pressure Filtration Units, Sartorius
Supplier: Sartorius
18052D 18053D 18056D 18058D 18068D 18072D 18075D
490013-570PK 190.34 CAD
490013-570 490013-572 490013-574 490013-576 76420-744 76420-746 76420-748
Sartolab® P20 and P20 Plus Pressure Filtration Units, Sartorius
Filters Membrane Filter Capsules
Ready to use units which, attached to a membrane pump or tube pump, quickly and reliably sterilise 100 ml to 5 litres of media and aqueous solutions.
  • Air vent with PTFE membrane
Cellulose acetate/PES membrane, P20 Plus with glass fibre prefilter; PC housing, sterile.
Outlet: Hose nipple with filling bell.
Non-toxic as confirmed with L929 fibrolast cells of mice and with MRC-5 lung cells of human embryonic origin.
Certifications: Pass the USP Plastics-Class VI-Test.
Packaging: Individually sterile bagged.