Infrared Moisture Analyzer, MA35, Sartorius
Supplier: Sartorius
MA35M-230N MA35M-115US MA35M-230N MA35M-115US
CA76412-334EA 3080 CAD
76412-332 CA76412-334 CA76412-332 76412-334
Infrared Moisture Analyzer, MA35, Sartorius
Analysers Moisture Analysers
This infrared moisture analyzer is designed for use in daily routine in-process control.
  • Uniform distribution of the heat rays using an integrated reflector
  • Heating of a sample by two metal tubular-shaped heating elements
  • Uniform distribution of the heat rays using an integrated reflector
Quick and reliable analysis of the material moisture of liquid, pasty and solid substances using the thermogravimetric method.
Certifications: FDA/HACCP regulations.