Vivaspin® 500 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
VS0101 VS0102 VS0111 VS0112 VS0121 VS0122 VS0131 VS0132 VS0141 VS0142 VS0152 VS0162 VS0172 VS0191 VS0192 VS0151 VS0161
CA76408-866EA 214.56 CAD
CA76408-866 CA76408-868 CA76408-870 CA76408-872 CA76408-874 CA76408-876 CA76408-878 CA76408-880 CA76408-882 CA76408-884 CA76408-886 CA76408-888 CA76408-890 CA76408-892 CA76408-894 CA76408-896 CA76408-898 CA76409-430 CA76409-432
Vivaspin® 500 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filter units offer a simple, one step procedure for sample concentration with volumes from 5 to 500 µl. They can effectively be used in fixed angle rotors accepting 2.2 ml centrifuge tubes.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
More Product Information