Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
VS2001 VS2002 VS2011 VS2012 VS2021 VS2022 VS2031 VS2032 VS2041 VS2042 VS2051 VS2052 VS2061 VS2062 VS2072 VS2091 VS2092
CA76409-066EA 268.02 CAD
CA76409-066 CA76409-068 CA76409-070 CA76409-072 CA76409-074 CA76409-076 CA76409-078 CA76409-080 CA76409-082 CA76409-084 CA76409-086 CA76409-088 CA76409-090 CA76409-092 CA76409-094 CA76409-096 CA76409-098 CA76409-100
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® 20 centrifugal concentrators offer increased volume flexibility and performance to the Vivaspin® potfolio. Vivaspin® 20 handles 5 to 20 ml in swing bucket rotors and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes. Vivaspin® 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows monitoring the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate.
  • High retentate recovery >90%
  • Vertical twin membrane for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • Polycarbonate body and filterate vessel, polypropylene concentrator cap with acetal or aluminum pressure head
For higher process flexibility, it is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer can be placed on the concentrator and spun another time to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure.
When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, it can be filled with up to 15 ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. 'Pressure-fugation' is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.