Weighing Tables for Micro and Analytical Balances, Sartorius
Supplier: Sartorius
YWT03 YWT04 YWT09
CA76413-088EA 3545.41 CAD
CA76413-088 CA76413-090 CA76413-092
Weighing Tables for Micro and Analytical Balances, Sartorius
Furniture Tables and Workbenches
High performance micro and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast stabilisation of the reading. The YWT09 weighing table features a sturdy frame with a granite stone slab insert.
  • Easy to clean surfaces
  • Fast stabilization times