Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories.
- Ideal for water and waste water analysis
- With dual silicon photodiodes detector
- Glove friendly touch-screen interface and keypad
AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation. Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP.
More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries. Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries.
Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards. Built-in test methods for stray light, noise and resolution are available to validate instrument performance further.
Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50 mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm Ø.
Certifications: TUV Complaince, CE certified.
Delivery information: Includes test tube holder for vials and test tube from 12 to 25 mm Ø, 10 mm square single-cell holder (Model AQ8100 only), AquaMate water analysis methods software, dust cover, external power supply, north america power cable (NEMA 5-15), european power cable (CEE 7/7), UK power cable (BS 1362).