VWR® Contour™ Fume Hood Base Cabinets
Supplier: VWR International
CFG-2084-V CFG-2085-V CFG-2086-V CFG-20860V CFG-2088-V CWG-2084-V CWG-2085-V CWG-2086-V CWG-2088-V
CFG-2084-VEA 2010.39 CAD
CFG-2084-V CFG-2085-V CFG-2086-V CFG-20860V CFG-2088-V CWG-2084-V CWG-2085-V CWG-2086-V CWG-2088-V
VWR® Contour™ Fume Hood Base Cabinets
Furniture Storage Cabinets
VWR® Venturi fume hood base cabinet.
VWR®/VFH-06/19-6 Steel cabinets (35" high) standing height steel and wood fume hood base cabinets are 35" high and a nominal 20" deep. ADA height steel fume hood base cabinets are 32" high and 20" deep; ADA height wood fume hood base cabinets are 32½" high and nominal 20" deep. Lengths as shown.
Order Now
SPECIFICATIONS
Environmentally Preferable