LED Sign Projectors, Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
VPL901 VPL902 VPL903 VPL904 VPL905 VPL906 VPL907 VPL908 VPL909 VPL910 VPL800 VPL911
76406-132EA 3524.16 CAD
76406-132 76406-136 76406-140 76406-144 76406-148 76406-152 76406-156 76406-160 76406-164 76406-168 76406-172 76406-176
LED Sign Projectors, Accuform®
Signs
LED Sign Projector is a perfect solution for generating signs in areas where conventional wall and floor signs won’t work.
  • Operates on 110 V with remote control
  • 3 ft. power cord
  • Has built-in mounting bracket
  • Lens is made of aluminum and glass with printed image
  • Additional message lens available
Viewing distance by light level:
Complete dark – 262-ft.; Dim light – 65-164-ft.; Bright light – 49-ft.
Certifications: CE and RoHS certified.
IP 65 rated for damp environments.