Zymo-Seq RiboFree™ Total RNA Library Kits, Zymo Research
Supplier: Zymo Research
R3000 R3001 D3008
76407-304EA 2021.15 CAD
76407-304 76407-306 76407-308
Zymo-Seq RiboFree™ Total RNA Library Kits, Zymo Research
Nucleic Acid Reagents Next Generation Sequencing Reagents
RNA-Seq made simple.
  • The fastest and easiest total RNA-seq library kit: Prepare stranded, ribofree libraries from total RNA in 3.5 hours
  • Compatible with any organism: Probe-free technology depletes rRNA and globin from any RNA source
  • The most accurate: Eliminate bias from rRNA depletion
Discover the most streamlined, stranded RNA library kit for sequencing total RNA from any biological sample with the Zymo-Seq RiboFree™ total RNA library kit. Capture any sample's full transcriptome (both coding and non-coding) with one simple RNA library prep kit. Also capture long-noncoding RNAs (lncRNA), intronic RNAs, nucleolar RNAs, and mRNAs with degraded poly(A) tails.
Ribosomal RNAs (rRNA) comprise approximately 90% of the total RNA and represent an obstacle to transcriptome enrichment of unique protein coding sequences. Zymo Research's RiboFree™ universal depletion effectively removes rRNA and overrepresented transcripts (e.g. globin). Without the use of probes that cause off-target digestion compared to popular rRNA removal kits.