Floor Stripe™ High Performance Marking Tapes, Accuform
Supplier: Accuform
ACCUFORM®
PTP102RD PTP103OR PTP102YL PTP103GN PTP103BU PTP103WT PTP103RD PTP102BK PTP103YL PTP102WT PTP102BU PTP102OR PTP103BK PTP102GN
76406-108RL 166.33 CAD
76406-108 76406-106 76406-104 76406-102 76406-088 76406-100 76406-086 76406-098 76406-084 76406-096 76406-082 76406-094 76406-092 76406-090
Floor Stripe™ High Performance Marking Tapes, Accuform
Tapes
Floor Stripe™ high performance marking tape provides visual guidance to mark and identify aisles, pathways, and designated locations.
  • Superior durability for marking in high-traffic locations
  • Acrylic adhesive backing provides high tack bond
  • Apply to clean, non-coated surfaces, including concrete
These tapes are constructed of low profile, polyester tape with high gloss overlaminate.