Slip-Gard™ Floor Marking Kits, Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
PSR523 PSR522 PSR521 PSR529
76405-586KT 122.48 CAD
76405-586 76405-588 76405-590 76405-592
Slip-Gard™ Floor Marking Kits, Accuform®
Signs
Slip-Gard™ Floor Marking Kits allows you to mark and identify specific areas around the workplace to keep clear of obstructions and debris to allow for general and emergency access.
  • Bold graphical symbol and messages convey the reason to keep area clear
  • Constructed of thick adhesive vinyl, subsurface printed, with clear laminate to protect the words and image
  • Non-slip, pebble-textured surface provides added durability to withstand heavy pedestrian traffic
  • Strong, permanent, self-adhesive underside has peel-off back liner
  • Adheres firmly to smooth, clean, dry floor surfaces
  • Kit covers a 36×24" area
Kit includes three pre-cut borders strips (two 2×36", one 2×24"), and one 17" round floor sign.