AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Supplier: Agilent
5062-2478 5062-2478 5190-9426
CA5062-2478EA 498.13 CAD
CA5062-2478 AG5062-2478 103584-582
AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Standards Biological Standards
AdvanceBio Amino Acid Analysis (AAA) includes proven reagents for amino acid derivatization, a ready-to-use amino acid standards kit, columns based on innovative Poroshell technology, and expert support.
  • Reliable results - high resolution separations delivered by efficient Poroshell particle morphology
  • Reduced costs - long column lifetimes from robust, high-pH resistant, chemically modified silica
  • Increased flexibility - compatibility with both HPLC and UHPLC systems via 2.7 m diameter particles
  • Quality control - AdvanceBio AAA columns are batch tested with amino acid standards to ensure quality
  • Automated online derivatization - with Agilent analytical injection systems
AdvanceBio AAA delivers fast, sensitive, and reproducible separations of amino acids in biological samples, protein hydrolysates, and cell culture media. Used alongside Agilent InfinityLab LC Series instruments, AdvanceBio AAA provides a complete solution for amino acid analysis.