LED Sign Projector Lens, Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
VPL901LENS VPL902LENS VPL903LENS VPL904LENS VPL905LENS VPL906LENS VPL907LENS VPL908LENS VPL909LENS VPL910LENS VPL800LENS VPL911LENS
76406-134EA 350.32 CAD
76406-134 76406-138 76406-142 76406-146 76406-150 76406-154 76406-158 76406-162 76406-166 76406-170 76406-174 76406-178
LED Sign Projector Lens, Accuform®
Signs
LED Sign Projector additional lenses are designed for use with LED Sign Projector (sold separately.)
  • Lens is made of aluminum and glass with printed image
Viewing distance by light level:
Complete dark – 262-ft.; Dim light – 65-164-ft.; Bright light – 49-ft.