Plastic Tube Brushes, Micronova
Supplier: Micronova
BRPL-25S36 BRPL-118 BRPL-1218 BRPL-1418 BRPL-3418 BRPL-32548
75993-596EA 114.39 CAD
75993-596 76415-970 76415-972 76415-974 76415-976 76415-978
Plastic Tube Brushes, Micronova
Brushes
These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
  • 100% polypropylene bristles and handle
  • Thin core and handles are flexible enough to bend through fittings
  • Available in a range of bristle diameters and handle lengths
Brushes feature narrow center cores that are flexible enough to pass through elbow tank fittings.
Certifications: FDA compliant materials.
Order Now
SPECIFICATIONS
Environmentally Preferable