Hand Held Tool and Covers, Micronova
Supplier: Micronova
HHT-06PP MZSM68-6S MZSM6-6NSIR
76342-622CS 134.81 CAD
76342-622 76342-624 76342-626 76342-628
Hand Held Tool and Covers, Micronova
Mops
The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
  • Ergonomic cleaning for small equipment and surfaces
  • Lightweight, forms to hand
  • Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
  • Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
  • PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
  • Covers compatible with alcohol and strong disinfectants and are launderable for reuse
  • Autoclavable
The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.