ESD Brushes, Menda
Supplier: Desco
35687 35688 35691 35692 35693 35694
76407-232EA 47.62 CAD
76407-232 76407-234 76407-236 76407-238 76407-240 76407-244
ESD Brushes, Menda
Brushes
ESD brushes are designed to help clean static sensitive equipment and components.
  • Ideal for removing dust, dirt, and foreign object debris from static sensitive components
  • Yellow bristles indicate dissipative brushes
  • Black bristles indicate conductive brushes
These ESD Brushes are designed to allow thorough cleaning of ESD sensitive assemblies without the danger of high charges generated by conventional brushes. Brushes are available with wither dissipative or conductive bristles.
Yellow dissipative bristles are highly resilient and abrasion resistant. They are hot stamped with the ESD protective symbol to indicate that they can be used in static sensitive applications. Black conductive bristles minimize static charge generation and remove electrostatic charges to ground. Conductive bristles are available in two styles, semi-fine and firm. Semi-fine bristles are ideal for chemical and electronics applications. Firm bristles are mainly for use with electronics, such as circuit boards.