Circular Conductive Connector Covers, Menda
Supplier: Desco
34208 34223
76407-218CS 937.29 CAD
76407-218 76407-220
Circular Conductive Connector Covers, Menda
Covers
Circular Conductive Connector Covers are designed to help keep connectors clean.
  • Non-corrosive
  • Free of carbon black particulate
  • Ideal for clean applications
A conductive cover for connectors. Ideal for clean applications because the cover does not leave black marks. Composite material provides a snug, secure fit which helps keep connectors clean and eliminates covers from falling off. Produces no visible corrosion when tested to the Federal Test Method 10C using Kovar test plates.