Analytical Balances, Entris II Basic Advanced, Sartorius
Supplier: Sartorius
BCA124-1S BCA124I-1S BCA224-1S BCA224I-1S BCA324I-1S BCA64-1S BCA64I-1S
CA76420-118EA 4348.27 CAD
CA76420-118 CA76420-120 CA76420-134 CA76420-136 CA76420-146 CA76420-168 CA76420-170
Analytical Balances, Entris II Basic Advanced, Sartorius
Balances and Scales
Entris® II Basic Advanced analytical balances feature isoCAL, intuitive graphic touch display and 13 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard-wearing PBT, stainless steel and glass. Integrated protection systems increase reliability of weighing results: 3 configurable levels determine valid weighing data and ensure only valid data is transferred to external devices.
  • Fast stabilisation time thanks to state-of-the-art weighing sensors
  • Best repeatability with
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable