Vivaspin® Turbo 15 RC, Sartorius
Supplier: Avantor
VS15TR12 VS15TR02 VS15TR21 VS15TR22 VS15TR31 VS15TR32 VS15TR42
CA76415-992PK 372.42 CAD
CA76415-992 CA76415-994 CA76415-996 CA76415-998 CA76416-000 CA76416-002 CA76416-004 CA76416-006 CA76416-008 CA76416-010
Vivaspin® Turbo 15 RC, Sartorius
Filters Centrifugal Filters
Vivaspin® Turbo 15 Regenerated Cellulose membrane, ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4 to 15 ml sample volumes.
  • Unique optimal membrane choice (PES and RC)
  • Complete recoveries
  • Fastest concentrations
  • Highest chemical resistance
  • Easy retentate collection
  • Graduated and markable
Vivaspin® Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs.
The Vivaspin® Turbo 15 RC concentrators are the regenerated cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 µl.