VWR® CryoPro® AF-VPS Auto-Fill Vapor Phase Systems
Supplier: VWR
VWR® CryoPro®
AF-VPSP-1 AF-VPSP-2PS AF-VPSP-1PS AF-VPSP-1CS AF-VPSP-3 AF-VPSP-2 AF-VPSP-3CS AF-VPSP-2CS AF-VPSP-3PS
76411-324EA 161403.41 CAD
76411-324 76411-314 76411-326 76411-330 76411-320 76411-322 76411-316 76411-328 76411-318
VWR® CryoPro® AF-VPS Auto-Fill Vapor Phase Systems
Cryopreservation Systems
These stainless steel, auto-fill vapor phase cryogenic storage systems are designed to maintain temperatures of samples well below the critical −135 °C (−211 °F) range, ensuring long-term cell viability.
  • Stainless steel fabrication
  • Long-term cell viability ensured
  • 4,8,12 13-2 mini racks
  • 14,30,58 13-2 standard racks
Samples are stored in an isolated inner aluminum tank, free of liquid, limiting cross-contamination concerns associated with sample storage in liquid.
The rotating storage tank has a reduced-size access opening with an easy-to-handle hinged lid, which limits the induction of harmful warm air during lid openings and reduces the system's consumption of liquid nitrogen. Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up, controller settings and an employee inservice by a factory representative.
A fully automatic controller and alarm package allows for convenient liquid level maintance. Sample security is monitored with a full array of alarms, including low liquid level, high liquid level, lid open, sensor failure, and power failure. Fully operational 3 to 5 day battery backup provides added security. Systems also include foldable skid-free step, five lockable casters, and one 1.8 m (6') armor-plated transfer hose.
Ordering information: Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up (which includes storage system start-up, controller settings, and personnel training by the manufacture).
Delivery information: All cardboard and plastic inventory systems include a 1.8 m (6') stainless steel transfer hose. AF-VPS 1 inventory systems include 14 standard racks, 4 mini racks, 182 cardboard or plastic 100-cell boxes, and 52 cardboard or plastic 25-cell boxes. AF-VPS 2 inventory systems include 30 standard racks, 8 mini racks, 390 cardboard or plastic 100-cell boxes, and 104 cardboard or plastic 25-cell boxes. AF-VPS 3 inventory systems include 58 standard racks, 12 mini racks, 754 cardboard or plastic 100-cell boxes, and 156 cardboard or plastic 25-cell boxes. All racks are stainless steel.
Caution: Static Evaporation Rates and Static Holding Times are nominal. Actual rates may be affected by the nature of the contents, atmospheric conditions, container history, and manufacturing tolerances.