Corning® Matrigel® Matrix-3D Plates, Corning
Supplier: Corning
356256 356257 356258 356259
76419-678CS 2148.03 CAD
76419-678 76419-680 76419-682 76419-684
Corning® Matrigel® Matrix-3D Plates, Corning
Microplates Cell Culture Plates
A Matrigel® pre-coated plate which can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids.
  • Available in 384 and 96-well ready-to-use formats
  • Easy and convenient as Matrigel® is pre-dispensed in each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded application assays
The Corning® Matrigel® matrix-3D plate is a high-throughput pre-coated format that contains Matrigel® and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids. Matrigel® Matrix is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Each lot is verified for intraplate consistency. Plates are tested and found negative for the presence of bacteria and fungi.