NPS® Steel Height Adjustable Science Lab Tables, Phenolic Top, National Public Seating
Supplier: National Public Seating
SLT3-2448P SLT3-2454P SLT3-2460P SLT3-2472P SLT3-3060P SLT3-3072P
76417-708EA 1798.58 CAD
76417-708 76417-720 76417-732 76417-744 76417-756 76417-768
NPS® Steel Height Adjustable Science Lab Tables, Phenolic Top, National Public Seating
Furniture Tables and Workbenches
Our ANSI-BIFMA Phenolic top steel adjustable height science table is an asset to every research lab.
  • Steel legs adjust smoothly and easily In 1" hole increments – from 22¼ to 37¼" height
  • Legs built with 1½" square tubing 14 gauge thickness
  • Legs built with "tamper-free" hex head screws and lock nuts
  • Weight capacity 500 lbs.
  • ¾" black phenolic top with matte finish
  • All legs come standard with simple-adjust glides
  • 10 year warranty
NPS Steel Science Tables are built to last and made for the toughest environments. 16 gauge steel frame apron and 14 gaugeMore Product Information