VWR® PCR® Cooler, 96-Well
Supplier: VWR
VWR®
120729V 120728V 120727V
76343-418PK 172.31 CAD
76343-418 76343-420 76343-416
VWR® PCR® Cooler, 96-Well
Coolers Benchtop Coolers
The VWR® PCR® Cooler is ideal for thermosensitive applications, PCR®, or cell-based assays, as well as gentle sample thawing and stopping of reactions.
  • Modern patented design cools faster in freezers and stays cold longer on the bench top
  • PCR® Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
  • When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
  • Cools industry standard format 0.2 ml and 0.5 ml PCR® tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
  • Better alternative than ice baths as dry incubation reduces the risk of contamination
  • Freeze cooler in an upright position enabling cold storage of tubes
  • Well design prohibits the freezing of samples
  • Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
  • Save space with compact footprint and stacking ability
  • The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
Perfect for transporting samples and reactions between preparation, thermal cycling and post-PCR® procedure areas.
Material: Polypropylene (Cooler); Polycarbonate (Lid).