Guardian 5000 Magnetic Hotplate Stirrers, OHAUS
Supplier: Ohaus
30500520 30500550 30500560
76417-274EA 1158.44 CAD
76417-274 76417-286 76417-290
Guardian 5000 Magnetic Hotplate Stirrers, OHAUS
Stirrers Magnetic Hotplate Stirrers
Reliable magnetic hotplate stirrers with exceptional safety features for everyday Use. These Guardian 5000 hotplate stirrers are designed for safety and simplicity in all research, academic and industrial applications.
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Easy to read LCD display
  • Designed for durability and reliability with SmartHousing™
The design ensures proper mixing and temperature control of the top plate or your sample when using the optional temperature probe. Heating models include a Single Point Calibration feature for multiple setpoints. Select from different models with various plate sizes for your everyday heating and stirring applications.
The early detection system uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
The bright LCD display includes large temperature and speed settings, intuitive icons and a prominent hot top indicator which are all designed to quickly and easily monitor performance from across the laboratory.
The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internal components and the angled control panel.
Corresponding magnetic stirrers (stirring only 76417-282) or hotplates (heating only 76417-278) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Certifications: Product Safety: TÜV SÜD certified to CAN/CSA C22.2 61010-1; CAN/CSA C22.2 61010- 2-010; CAN/CSA C22.2 61010-2-051; UL 61010-1; UL 61010-2-010; UL 61010-2-051
Electromagnetic Compatibility: Canada ICES-003 Class A; FCC Part 15 Class A
Compliance Mark: TÜV SÜD
Accessories information: Built-in support rod holder, PTFE stir-bar.
Ordering information: Two year manufacturer's warranty.