Brady® Spill Response Plus Chemical Absorbent Pads, Brady Worldwide
Supplier: Brady Worldwide
SRPH144 SRPH200
76417-796CS 231.51 CAD
76417-796 76417-798 76417-800 76417-802
Brady® Spill Response Plus Chemical Absorbent Pads, Brady Worldwide
Spill Control Materials
The universal, multi-purpose absorbent is made to handle virtually any liquid. It is designed to be highly absorbent, durable, and waste reducing.
  • Surfactant-treated; brightly colored absorbent picks up nearly any leak or spill
  • Heavyweight design for continuous leaks and larger spills
  • Small, convenient size is ideal for employees to fold and carry in pockets for quick spill response
  • Non-perforated roll with no disruption in the material; improves absorbency and tear-resistance in high traffic areas
  • Compact sizes
Spill response plus chemical absorbent pads support compliance with UN, IATA, ICAO, and parcel carrier requirements for shipping diagnostic specimens and infectious substances. These dimpled, low-linting, and environmentally friendly pads are also fire-resistant and highly durable (15 to 27 lbs.).
Certifications: EPA 40 CFR 112.7(c)(1)(vii);OSHA 29 CFR 1910.120(j)(1)(vii);OSHA 29 CFR 1910.22(a)(2)