VWR® Low-Profile Refrigerated Circulating Baths
Supplier: Polyscience
AP7LR-20-V11B AP7LR-20-V12E
76415-932EA 11240.42 CAD
76415-932 76415-934
VWR® Low-Profile Refrigerated Circulating Baths
Circulating Thermostats Closed Circulator Systems Cooling Circulators
VWR® Refrigerated circulating baths offer precise temperature control, at temperature ranges from –20 to 200 °C.
  • OpenMode time time/temperature programming (no restrictions on number of programs or steps)
  • Intuitive 10.9 cm (4.3") SmartTouch display
  • 11 languages: English, French, German, Spanish, Chinese, Portuguese, Russian, Hindi, Arabic, Italian and Korean
  • Variable speed pump with open and closed-loop external circulation capability
The Advanced Series Controllers offer Swivel 180™ technology, which allows the user to rotate the controllerMore Product Information