Blue Phantom® Gen II Femoral Line and Regional Anesthesia Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPP-136
470327-384EA 9835.39 CAD
470327-384
Blue Phantom® Gen II Femoral Line and Regional Anesthesia Ultrasound Trainer
Models Human Biology Models
Matches the acoustic characteristics of real tissue.
  • Inject simulated anesthetics
  • Superb ultrasound imaging characteristics
  • Self-healing tissue
  • Use for blind, or ultrasound guided insertion
  • Positive fluid flow in the vessels provides users with immediate feedback when vessels are accessed
  • Use any ultrasound imaging system
Blue Phantom®'s Generation II Femoral lower torso ultrasound guided regional anesthesia and central venous access training model is an excellent tool to train users to develop and practice the skills necessary to gain proficiency in using ultrasound for regional anesthesia procedures of the femoral nerve as well as guide catheter insertions in the femoral artery and vein.
Built upon the feedback received from customers of the first generation femoral model, Blue Phantom® has developed the most realistic Femoral Ultrasound trainer available in the market today. Using Blue Phantom® simulated human tissue, this very realistic and ultra-durable ultrasound training phantom is an excellent choice for training clinicians in the psychomotor skills associated with ultrasound guided regional anesthesia as well as vascular access procedures.
These ultrasound imaging skills include; using ultrasound system controls, transducer positioning and movement, recognition of nerves, arterial and venous anatomy, using ultrasound to target the femoral nerves for regional anesthesia procedures, guide needles to blood vessels for cannulation, and performing vascular access procedures.
Comprised entirely of simulex self-healing tissue, users can access vessels on both limbs resulting in twice the number of access sites compared to the original model. With the innovative new hand pump system, the femoral artery no longer collapses creating our most realistic anatomy yet.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.