Blue Phantom® Gen II Regional Anesthesia and Central Line Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
BPHNB670-AP BPHNB670-HP BPHNB673
470327-376EA 19889.29 CAD
470327-376 470327-386 470327-532
Blue Phantom® Gen II Regional Anesthesia and Central Line Ultrasound Trainer
Models Human Biology Models
Cast from a live patient.
  • Matches the acoustic properties of real tissue
  • Pulse using automatic pump of hand bulb options
  • Self-healing tissue
  • Use with any ultrasound system
Blue Phantom®'s new ultrasound central line and regional anesthesia training model is the most realistic central line and nerve block simulator available anywhere. Cast from a live human, innovative Life Cast™ modeling approach provides the most realistic external landmarks while the internal anatomy is based on digital human files. This ultra-durable mannequin incorporates all ofMore Product Information