Blue Phantom® Central Line Ultrasound Trainers
Supplier: ELEVATE HEALTHCARE, INC. SE
BPH660-AP BPH660-HP BPH665-C-HP BPH662-B BPH667-C BPH663
470327-380EA 14105.29 CAD
470327-380 470327-382 470327-390 470327-394 470327-470 470327-536 470327-538
Blue Phantom® Central Line Ultrasound Trainers
Models Human Biology Models
Superior realism, reliability, and durability.
  • Self-healing tissue offers users tremendous durability
  • Anatomically correct; constructed from a digital human file
  • Veins are compressible while the arteries remain uncompressed
  • Easy to refill vessels using Quick fill™ port
Blue Phantom® is proud to introduce the most realistic ultrasound central line mannequin available anywhere. Constructed utilizing the innovative Life Cast™ modeling approach, the external landmarks were cast from a live human while the internal anatomy is based on digital human files.More Product Information