BioFit Bridgeport Cleanroom Swivel Chairs, ISO 3
Supplier: BioFit
Bridgeport
BTALRTXX3C540 BTSHHTXX3C540 BTAMRTXX3C540 BTSLRTXX3C540 BTSMHRXX3C540
76412-134EA 3033.13 CAD
76412-134 76412-154 76412-146 76412-108 76412-130
BioFit Bridgeport Cleanroom Swivel Chairs, ISO 3
Chairs
Bridgeport Series chairs boast a number of features and benefits that make them a popular choice for laboratory, healthcare, education, technical, industrial and office workspaces. The ergonomic design enhances user comfort and productivity when sitting for prolonged periods.
  • Concave ergonomic seat
  • Nimble, trapezoid-shaped backrest which provides lumbar support
  • Easily adjusted controls to meet the needs of multiple shift users
  • Easy to effectively clean and disinfect
Seating solutions are crafted with user comfort, health and wellbeing in mind. Products meet or exceed rigorous industry standards and nearly all carry a 13-year warranty with a lifetime warranty on the pneumatic mechanisms.
Products are renowned for proven performance and reliability, backed by a history of over 75 years improving the workplace experience by providing ergonomic seating for laboratory, healthcare, technology, education, industrial, office and custom applications.
Certifications: LEED® compliant for use in commercial interiors; complies with ANSI/BIFMA X5.1 product safety and performance standards.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.