Anti-Rabbit CyDye™ 700 NIR-Labelled Secondary Antibody
Supplier: Cytiva
29360786 29360787
CA76325-080EA 193.11 CAD
CA76325-080 CA76325-082
Anti-Rabbit CyDye™ 700 NIR-Labelled Secondary Antibody
Antibodies
Amersham CyDye NIR-labeled secondary antibodies enable quantitation of Western blots with a strong signal-to-noise ratio. Mulitplexed experiments allow for the detection of two proteins on a single blot. The use of total lane signals to normalize target protein data yield robust and reliable results.
  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation
More Product Information