Biotin-20-dUTP, Biotium
Supplier: Biotium
40030
40030-1
89139-094EA
571.63
CAD
89139-094
89139-096
Biotin-20-dUTP, Biotium
Protein Linking and Labeling Systems
Biotin-20-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling. Biotin-dUTP conjugates are available wtih 11-carbon, 16-carbon, or 20-carbon linker lengths.
- 20-carbon linker between biotin and dUTP
- Available in solid or solution form
- Solid contains lyophilized TE buffer
- Solution is 1 mM in Tris-HCl buffer pH 7.5
In general, the shorter the linker, the more efficiently the biotin-dUTP is incorporated into DNA by DNA polymerases. On the other hand, a longer linker should facilitate interaction between biotin and avidin or streptavidin.