Biotin-11-dUTP, Biotium
Supplier: Biotium
40029
40029-1
89139-090EA
521.1
CAD
89139-090
89139-092
Biotin-11-dUTP, Biotium
Protein Linking and Labeling Systems
Biotin-11-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling.
- 11-carbon linker between biotin and dUTP
- Available in solid or solution form
- Solid contains lyophilized TE buffer
- Solution is 1 mM in Tris-HCl buffer pH 7.5
Biotin-dUTP conjugates are available wtih 11-carbon, 16-carbon, or 20-carbon linker lengths. In general, the shorter the linker, the more efficiently the biotin-dUTP is incorporated into DNA by DNA polymerases. On the other hand, a longer linker should facilitate interaction between biotin and avidin or streptavidin.