Test Strips, Water Hardness, Dosatest, VWR Chemicals BDH®
Supplier: VWR International
91201.AVA 91220.AVA
BDH85442.601EA 142.46 CAD
BDH85442.601 BDH85443.601
Test Strips, Water Hardness, Dosatest, VWR Chemicals BDH®
Environmental Test Kits
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor water hardness, especially suitable for boiler testing, aquaria and laundries. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
1 mmol/litre =10 °f = 5.60 °d= 7,022 °e
1 mmol/litre= 100.09 mg/l CaCO₃
Packaging: Container with 100 test strips 6 x 95 mm