Test Strips, Nitrate/Nitrite, Dosatest, VWR Chemicals BDH®
Supplier: Avantor
91313.200131.1
BDH85439.601EA 152.81 CAD
BDH85439.601
Test Strips, Nitrate/Nitrite, Dosatest, VWR Chemicals BDH®
Environmental Test Kits
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6 x 95 mm.