Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Supplier: VWR International
BDH85433.601 BDH85434.601 BDH85435.601
BDH85433.601EA 135.07 CAD
BDH85433.601 BDH85434.601 BDH85435.601
Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Environmental Test Kits
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.