HyClone™ Characterized Fetal Bovine Serum, CA Origin, HyClone products (Cytiva)
Supplier: Cytiva
3039603IH25-40 3039603IR25-40 3039603IR30-45
CA76324-900EA 617 CAD
CA76324-900 CA76324-898 CA76324-932
HyClone™ Characterized Fetal Bovine Serum, CA Origin, HyClone products (Cytiva)
Sera (Serum)
A cost-efficient alternative to defined FBS, this high-quality Canadian serum is well-characterized.
  • Characterized: assayed for a variety of critical components and biochemical characteristics
  • Canadian origin: sourced in Canada and processed in the US with complete traceability
  • Endotoxin-tested: certificate of analysis confirms ≤25 EU/ml endotoxin
  • Tripled-filtered: serial 100 nm pore size-rated filterings for reliable microbial depletion
γ-irradiated and heat-inactivated FBS options also available.
Certifications: Broad virus panel testing according toin accordance with 9 CFR 113.53