HyClone™ Defined Fetal Bovine Serum (FBS), US Origin, HyClone products (Cytiva)
Supplier: Cytiva
SH30070.03IH25-40 3007003IR30-45
CA76324-860EA 1353 CAD
CA76324-860 CA76324-920 CA76324-858 CA76324-918 CA82013-566
HyClone™ Defined Fetal Bovine Serum (FBS), US Origin, HyClone products (Cytiva)
Sera (Serum)
Highest quality FBS, HyClone US-origin defined FBS is 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profile.
  • Well-defined: assay results for over 50 components, with additional testing available
  • US origin: sourced and processed in the United States with complete traceability to origin
  • Ultra-low endotoxin: ≤10 EU/ml endotoxin for reduced research and production risk
  • Sterile-filtered: serial 40 nm filter process removes up to 8 logs viral challenge
  • No mycoplasma is present
γ-irradiated and heat-inactivatedMore Product Information