PIG® High-Visibility Safety Floor Runners, New Pig
Supplier: New Pig
GRPSB36200-BK GRPSB36201-BK GRPSB36206-BK
76378-334RL 613.67 CAD
76378-334 76378-336 76378-338
PIG® High-Visibility Safety Floor Runners, New Pig
Spill Control Materials
Provide a no-slip path in hazardous walking areas with this super-safe, adhesive-backed absorbent runner — its hi-viz yellow safety edges can't be missed.
  • Highly visible yellow and black marking is designed to meet ANSI standard for floor markings and help you comply with OSHA 1910.22 (walking surfaces standard)
  • Durable printed surface withstands industrial wear and tear; even forklift traffic
  • Mat is highly absorbent, quick-drying, very low profile, extremely durable and effortless to maintain
  • Easy to use and customize: roll out continuous coverage with no gaps, ripples or overlaps that cause slips and trips; create custom-sized floor runners, even around corners
  • Proprietary adhesive sticks to the floor with a super-tight grip, but peels up easily to replace when mat shows signs of wear
  • Sticks to most commercial floor surfaces including concrete, VCT, linoleum, ceramic tile, quarry tile and laminate flooring
  • Easy to clean: you can sweep, vacuum, mop, use your shop/wet vac or run your floor scrubber and it won’t budge
  • Absorbs a wide variety of liquids including water, oil and grease; leakproof backing prevents liquid from soaking through to the floor
Tapes and paints are easy to see but do nothing to create a safer floor surface. The PIG High-Visibility Safety Floor Runner is the only floor safety solution that combines a high-traction walking surface with a hi-viz safety edge pattern.
Adhesive backing keeps mat exactly where you put it with no shifting, bunching up or flipping over; traps dirt, salt and moisture to keep it from being tracked around.
The thinnest commercial floor mat you can buy: no rippled edges to obstruct doors or create tripping hazards; ADA compliant for wheelchairs and walkers; carts and pallet jacks roll right over it.
Certifications: Tested and Certified by the National Floor Safety Institute (NFSI): when used as part of your floor safety program, high-traction surfaces such as PIG Grippy Floor Mat have been proven to reduce slip-and-fall claims by as much as 90%.
Ordering information: Installation Tool and Cutting Guide is a convenient, pre-scored cutting board that prevents damage to floors while it helps you make clean, accurate cuts, perfect corners and 45° angles. Safety Cutting Knife with recessed blade and protective edges lets you make quick, clean cuts and avoid injuries.