Avantor® ACE® Excel® Oligo, Analytical, HPLC/UHPLC Columns
Supplier: Avantor
EXL-1115-0502 EXL-1115-1002 EXL-1115-1046 EXL-1115-1502 EXL-1115-1546 EXL-1715-0502 EXL-1715-1002 EXL-1P15-GD2U EXL-1P15-GD4U
76415-432EA 2110.56 CAD
76415-432 76415-434 76415-436 76415-438 76415-440 76415-442 76415-444 76415-446 76415-448 76415-450
Avantor® ACE® Excel® Oligo, Analytical, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Oligo is a unique encapsulated stationary phase for high resolution ion-pair reversed-phase analysis of oligonucleotides. These high pH stable stainless steel columns are compatible with all buffers and mobile phases typically used for oligonucletide analysis by LC-UV and LC-MS, including TEAA and HFIP.
  • Ultra-inert, base deactivated silica for exceptional reproducibility and peak shape
  • Excellent column lifetime under oligonucleotide testing conditions
  • Ultra-low bleed for excellent LC/MS compatibility
High efficiency 1.7 and 3.0 µm particle sizes for high resolution oligonucleotide separations.
These columns are made of stainless steel.