Elmadry TD Dryers, Hot or Cold Air, Elma
Supplier: Elma
1082664 1082662 1082666
76326-622EA 2131.17 CAD
76326-622 76326-614 76326-670
Elmadry TD Dryers, Hot or Cold Air, Elma
Ultrasonic Cleaners
Elmadry TD dryers are used to safely dry small parts after ultrasonic cleaning and rinsing. The Elma basket is transferred directly from the ultrasonic cleaner to the dryer. The dryer can be set to either hot air drying (70 °C/158 °F) or cold air drying.
  • Hot air (70 °C) and cold air blower for drying sensitive parts
  • Simple and fast operation
  • Safety shutdown upon reaching the limit temperature
  • Three dryer sizes for Elma basket sizes 30, 120 and 300
  • Dust filter
Elmadry TD drying units are available in three sizes (30, 120 and 300) and are designed for use in production, workshop and service areas. For spotless drying after ultrasonic cleaning, the basket full of parts is first rinsed in tap water and then dipped into a bath of deionized or distilled water. The basket with the cleaned parts is then conveniently placed in the Elmadry TD drying unit.
The hot air function is activated while the cover is on. The cover has ventilation slots, ensuring that maximum air turbulence is produced and the moist air is conducted to the outside. At the same time, the cover reduces energy consumption. The dust filter on the underside is replaceable and prevents re-soiling of the cleaned parts by the dust in the room air.
Fitted with a cold and hot air blower, sensitive parts can be dried gently. Cold air operation can be used to rapidly cool dried parts.
Dependable, state-of-the-art Elma dryers are made in Germany.
Certifications: CE, ISO9001, ISO13485
Delivery information: All models supplied with cover, power cable, and dust filter.