Cloth Tapes, DUKAL™ Corporation
Supplier: Dukal
C15 C210 C50
76407-970BX 70.18 CAD
76407-970 76407-972 76407-974 76407-976
Cloth Tapes, DUKAL™ Corporation
First Aid Supplies First Aid Kit Components
The perfect amount of tape for securing dressings, I.V. sites, or tubing.
  • Patient specific, single use rolls
  • Contributes to the safe and sanitary treatment of all patients
  • Designed for bilateral tearing without use of scissors
  • No natural rubber latex
Durable cloth tape is designed for strength, with a gentle adhesive that will leave minimal residue on the skin.
Order Now
SPECIFICATIONS
Environmentally Preferable