Mouse Recombinant EGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-53AF-100UG 200-53AF-1MG 200-53AF-500UG 200-53AF-5MG
10788-228EA 151.36 CAD
10788-228 10788-230 10788-232 10821-746
Mouse Recombinant EGF (from E. coli)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
  • High quality
  • Low endotoxin
  • Affordable