Human Recombinant EGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-26AF-100UG
100-26AF-1MG
100-26AF-500UG
100-26AF-5MG
10787-468EA
151.79
CAD
10787-468
10787-470
10787-472
10821-742
Human Recombinant EGF (from E. coli)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.
- High quality
- Low endotoxin
- Affordable