BINDER −86 °C Ultra-Low Temperature Upright Freezers, UF V Models
Supplier: Binder
9020-0351 9020-0352 9020-0353 9020-0354 9020-0454 9020-0455
76413-536EA 35673.46 CAD
76413-536 76413-538 76413-540 76413-542 76836-760 76836-762
BINDER −86 °C Ultra-Low Temperature Upright Freezers, UF V Models
Freezers Ultra Low Temperature Freezers
These ultra low temperature freezers ensure the safe storage of samples at –80 °C. These combine outstanding environmental friendliness with low energy consumption, convenient operation and an individual safety concept.
  • Range of temperature controller –86 to –40 °C
  • Lowest energy consumption in its class
  • Powerful cascade compressor cooling unit
  • Environmentally friendly refrigerants R-290 and R-170
  • Efficient thermal insulation with vacuum insulation panels
  • Zero-voltage alarm contact for in-house alarm system
  • Features Ethernet interface and elapsed time
More Product Information