Mouse Recombinant IL-36 gamma (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Interleukin 36 gamma (IL-36 ɣ) is a member of the interleukin 1 (IL-1) cytokine family and protects against pathogens in the skin, lung, and stomach epithelial barriers. IL-36 ɣ binds the interleukin-1 receptor accessory protein (IL-1RAcP) and the orphan IL-1R-related protein 2 (IL-1Rrp2) receptors to activate NF-kB and MAP kinase signaling pathways, resulting in the induced production of inflammatory cytokines and chemokines.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.