Human Recombinant FGF-6 (from E. coli)
Supplier: Fujifilm Irvine Scientific
100-201AF-5UG
100-201AF-25UG
100-201AF-1MG
100-201AF-100UG
100-201AF-500UG
76050-902EA
142.6
CAD
76050-902
76050-904
76050-906
76050-908
76050-910
77660-736
77660-738
Human Recombinant FGF-6 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.
- High quality
- Low endotoxin
- Affordable