Undercounter Lab Glassware Washers, Model PG8593, Miele
Supplier: Miele
62859301CDN 99999527
CA75806-556EA 26088.88 CAD
CA75806-556 CA77445-684
Undercounter Lab Glassware Washers, Model PG8593, Miele
Glassware Washers
Glassware washer with liquid dispensing & DryPlus drying. Perf./cycle, e.g., 128 laboratory flasks.
  • Efficient use of resources - variable speed heater pump
  • Safety through monitoring - wash pressure and spray arm monitoring
Efficient hot air drying - DryPlus Total security - link to process documentation possible DryPlus for perfect drying results: The hot air drying feature DryPlus is the perfect solution for hollow body instruments.
The upstream H13 HEPA filter ensures optimum particle separation from the drying air. The filter can be easily accessed via a cover in the front of the base area. Short batch times: The Miele Professional laboratory glassware washer clean even demanding batches quickly and reliably. External detergent pump DOS K85 or DOS K85/1 must be ordered.
Certifications: EN 61010-1, EN 601010-1, CSA approved, CE-marked.
Ordering information: Baskets and accessories are not included when purchasing just the glassware washer, these must be ordered separately. Please contact your Avantor Sales representative/technical services for assistance with the product configuration. Packages including glassware washer, accessories and chemicals are also available please see details below.