Rat Recombinant M-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-33AF-10UG 300-33AF-100UG 300-33AF-1MG 300-33AF-500UG
10821-728EA 157.68 CAD
10821-728 10821-730 10821-732 10821-734 10822-274
Rat Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy.
  • High quality
  • Low endotoxin
  • Affordable